Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Excelta™ Precision Tweezers with Straight Strong Medium Tips
Precision Tweezers feature strong tips for long life.
Supplier: Excelta™ AC
Description
Strong tips for long life
- Anti-magnetic stainless steel or Carbon Steel
Specifications
Straight, Strong, Medium Fine | |
4.25 in. | |
10.7 cm | |
1 Ea. | |
0.03 cm | |
0.09 in. | |
0.0236 in. | |
0.059 cm | |
0.348 mm | |
Straight |
AC | |
Carbon Steel | |
Tweezers, Electronic | |
0.015 in. | |
High | |
0.32 in. | |
0.8 cm | |
0.0137 in. | |
1 Ea. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction