Learn More
Excelta™ Excelta™ Reverse-Action Stainless Steel Tweezers
Tweezers designed to hold objects without finger strain
Manufacturer:  Excelta™ DN5SN
 View more versions of this product
Catalog No. 18-100-841
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • AISI 303 anti-magnetic, high-precision stainless-steel
  • Closed at rest, press to open
  • Ideal for holding any small items for extended periods of time
Specifications
Specifications
Straight Tapered, Very Fine
Stainless Steel
Tweezers, Reverse Action
Holding any small items for extended periods of time
2.54 cm
0.32 in.
0.81 cm
0.01 in.
4.25 in.
108 mm
Each
1 in.
Extreme
0.01 in.
0.025 cm