Learn More
Excelta™ Small Parts Handling Tweezers
Manufacturer:  Excelta CorporationSupplier Diversity Partner 41
 View more versions of this product
Catalog No. 17-467-259
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Small Curved Flat Tip
Carbon Steel
Tweezers
It has two grooves in the tips which allow pickup from flat surfaces, parts to be handled can be no more than 0.01 in. thick
0.01 in.
4.5 in.
11.43cm
Each
Three Star High