Learn More
Excelta™ Excelta™ Ultra Precision (Five Star) Black Replaceable Ceramic Pointed Tweezer - ESD Safe
Zirconia ceramic has working temperature of over 2000degreeF
Manufacturer:  Excelta™ 3CCEESD
Catalog No. 18-100-909
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Straight fine Static dissipative tip
  • Used with very high magnification
  • Non-magnetic and chemically inert
Specifications
Specifications
Straight, Fine
Zirconia Ceramic
Tweezers, Straight Fine Point ESD Safe
1.5 mm
0.375 in.
5.25 in.
130 mm
0.06 in.
Ultra
9.5 mm