Learn More
Metrohm USA EXCHANGE UNIT 20ML
Supplier: Metrohm USA 63026220
Exchange unit with integrated data chip with 20 mL glass cylinder and light protection. PCTFE/PTFE flat stopcock, FEP tubing connection, anti-diffusion buret tip and standard amber glass reagent bottle.