Learn More
YSI™ Exchangeable Unit with Calibration Certificate
Intelligent exchangeable heads for the piston burette TITRONIC™ 500 and the titrators TitroLine™ 7000, 7500, 7750 und 7800.
$1505.52
Specifications
Type Intelligent Exchangeable Unit for TitroLine 7000+ and TITRONIC 500
For Use With (Equipment) TitroLine 7000+ and TITRONIC 500
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Sample Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Sample Price Quantity & Availability  
15-177-867
View Documents
YSI™
285230720
10 mL Each for $1,505.52
Add to cart
15-177-868
View Documents
YSI™
285230730
20 mL Each for $1,505.52
Add to cart
15-177-869
View Documents
YSI™
285230740
50 mL Each for $1,505.52
Add to cart
Description
Description
Intelligent exchangeable heads for the piston burette TITRONIC™ 500 and the titrators TitroLine™ 7000, 7500, 7750 und 7800.
  • Size options of 5, 10, 20 und 50 ml
  • Compact, space saving footprint
  • Easy to exchange
  • All relevant reagent and unit data are stored in the integrated RFID-chip including:
    - Burette size (ml)
    - Reagent/titrant name
    - Titrant concentration or titer value of solution
    - Date of manufacture or expiration date of the reagent.
    ISO 8655 Certified (“C” certificate included)
Scope of delivery:
5, 10 , 20 or 50 ml burette, with integrated chip for reagent data, with 1 L brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tip
Specifications
Specifications
Intelligent Exchangeable Unit for TitroLine 7000+ and TITRONIC 500
TitroLine 7000+ and TITRONIC 500