Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXD3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549011

Catalog No. PA549011

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Possesses 3 ft.-5 ft. exoribonuclease activity. Required for 3 ft.-end trimming of AGO1-bound miRNAs.
Specifications

Specifications

EXD3
Polyclonal
PBS with 0.09% sodium azide
Q9NX53
Exonuclease mut-7 homolog, 31--, Exonuclease 3′-5′ domain-containing protein 3, EXD3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54932
Western Blot
Unconjugated
EXD3
Liquid
EXD3
KLH conjugated synthetic peptide between 340-367 amino acids from the Central region of human EXD3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit