Learn More
Exel International Huber Needles
Supplier: Exel International 26901
Description
- Sterile
- Non-coring
- 316L stainless steel construction
- Available in both straight and 90 degree angles
- Needle volume of +0.5mL
- Polypropylene hubs, color-coded according to gauge for easy identification