Learn More
Exel International Huber Needles
Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
14-841-69
View Documents
Exel International
26901
N/A Huber Needle Rt. Angle (Non-Coring) 20G x 0.75 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
14-841-72
View Documents
Exel International
26904
N/A Huber Needle Rt. Angle (Non-Coring) 22G x 1 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
14-841-81
View Documents
Exel International
26923
N/A Huber Needle Rt. Angle (Non-Coring) 20G x 1.5 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
  • Sterile
  • Non-coring
  • 316L stainless steel construction
  • Available in both straight and 90 degree angles
  • Needle volume of +0.5mL
  • Polypropylene hubs, color-coded according to gauge for easy identification