Exel International Multi-Sample Blood Draw Needles

Multi-sample needles permit several samples to be taken with a single puncture

Manufacturer: EXEL INTERNATIONAL 26503

 View more versions of this product

Catalog No. 14-840-05

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Color Code Green
    For Use With (Application) Provides several samples to be taken with a single puncture, make penetration painless for patients and provides easy connection to rubber stoppers
    Gauge 21g
    Includes Sharp and smooth edge
    Latex Free Yes
    Length (English) 1 in.
    Length (Metric) 2.5cm
    Product Type Multi Sample Blood Draw Needle
    Sterile Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit