Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exel International Safety AV Fistula
Supplier: Exel International 27781
Specifications
Siliconized | |
Reduces possibility of accidental needle sticks, allow maximum blood flow | |
Yes | |
Secure Touch™ Av Fistula | |
Ultra-sharp |
15 G | |
Backeye, 12 in. tubing, clamp and beige fixed wing, ultra-thin wall needle, luer cap | |
1 in. | |
50 Box |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction