Exel International Super-ject™ Dental Needles

Plastic hub design allows for greater needle flexibility and lowers chances of needle breakage

Manufacturer: EXEL INTERNATIONAL 26558

 View all options for this product

Catalog No. 14-840-14


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Gauge 27 ga
    Length (English) Needle 0.8 in.
    Length (Metric) Needle 21mm
    Type Dental Needle
    Color Yellow
    For Use With (Application) For greater needle flexibility and lowers chances of needle breakage
    Material (Hub) Plastic

    • Red dot on the needle hub marks the needle bevel position, even when the needle point is not visible
    • Dental needle production is computer controlled in all processes from manufacture to inspection
    • Siliconized cannula
    • Needles are sterilized by Ethylene Oxide
    • Dental needle hubs and boxes are color-coded by gauge for easier identification
    • Self-tapping ribs placed on the outside of the hub, and inside threads, make for ease of use
    • Larger cannula lumen (.0055 in.)

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit