Learn More
Exel International Tuberculin Syringes
Specifications
Certifications/Compliance Pyrogen Free
Type Tuberculin Syringe
Volume (Metric) 1 mL
For Use With (Application) Positive plunger stop to prevent spillage when drawing, smooth plunger action for greater filling accuracy, non-toxic, pyrogen free
Latex Free Yes
View More Specs
Products 5
Catalog Number Mfr. No. Connection Type Length (English) Length (Metric) Needle Gauge Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type Length (English) Length (Metric) Needle Gauge Price Quantity & Availability  
14-840-50
View Documents
Exel International
26040
Removable 0.5 in. 1.3 cm 27 G
Pack of 100 for $61.22
Case of 10 PK for $508.07
N/A
Due to product restrictions, please to purchase or view availability for this product.
14-840-51
View Documents
Exel International
26043
Removable 0.5 in. 1.3 cm 26 G
Pack of 100 for $61.22
Case of 10 PK for $508.07
N/A
Due to product restrictions, please to purchase or view availability for this product.
14-841-34
View Documents
Exel International
26046
Fixed 0.62 in. 1.6 cm 25 G
Pack of 100 for $76.10
Case of 5 PK for $325.18
N/A
Due to product restrictions, please to purchase or view availability for this product.
14-840-53
View Documents
Exel International
26048
Luer Tip No Needle Included No Needle Included No Needle Included
Pack of 100 for $47.02
Case of 10 PK for $401.63
N/A
Due to product restrictions, please to purchase or view availability for this product.
14-840-52
View Documents
Exel International
26044
Removable 0.62 in. 1.6 cm 25 G
Pack of 100 for $61.22
Case of 10 PK for $508.07
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Latex free
  • Sterile
  • Non-toxic
  • Non pyrogenic
  • Reduced dead space for reduced medication waste
  • Smooth plunger action for greater filling accuracy
Specifications
Specifications
Pyrogen Free
1 mL
Yes
Tuberculin Syringe
Positive plunger stop to prevent spillage when drawing, smooth plunger action for greater filling accuracy, non-toxic, pyrogen free
Sterile