Learn More
Exel International ZorBand™ Pressure Bandages
Manufacturer:  Exel International 26834
 View more versions of this product
Catalog No. 14-840-28
Add to cart
Alert:
Caution: This product contains natural rubber latex which may cause allergic reaction.
Description
Description
  • Sterile and individually packaged
  • More convenient than traditional dressings
  • Reduces direct pressure time by over 40%
  • Absorbent cellulose pad expands when contacted by blood
  • Swelling action of pad creates a pressure compress
  • Firmly applies pressure to puncture
  • Tightly binds to patient's skin to guard from leakage and maintain pressure
Specifications
Specifications
Cellulose
Sterile
Large
Pressure Bandage
100 Pk.