Learn More
Exendin-4 Mouse anti-Human, Alexa Fluor™ 594, Clone: 24, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223579AF594
Catalog No. NP22379A94
Add to cart
Description
Description
Exendin-4 Monoclonal antibody specifically detects Exendin-4 in Human samples. It is validated for ELISA.
Specifications
Specifications
Exendin-4
24
ELISA
Exendin4
Synthetic exendin-4 adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells.
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human