Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exendin-4 Mouse anti-Human, Clone: 24, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Exendin-4 |
---|---|
Clone | 24 |
Concentration | 1 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22357901
|
Novus Biologicals
NBP2235790.1ML |
0.1 mL |
Each for $436.00
|
|
NB019343
|
Novus Biologicals
NBP2235790.025ML |
0.025 mL |
Each for $176.85
|
|
Description
Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.Specifications
Exendin-4 | |
1 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
Synthetic exendin-4 adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4C. Do not freeze. |
24 | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Human | |
Exendin4 | |
IgG1 κ | |
Protein A or G purified | |
Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title