Learn More
BioworldSupplier Diversity Partner EXENDIN-4 (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220600761
Exendin-4 Recombinant is a novel 39-amino acids peptide having a molecular mass of 4186.7 Dalton, which shares 53%, sequence homology with GLP-17-36 amide and interacts with the same membrane receptor. The Exendin-4 is purified by proprietary chromatographic techniques. Appearance:Sterile Filtered White lyophilized (freeze-dried) powder. Purity: Greater than 95.0%. MW 4186.7 Size 20ug
Catalog No. 50-253-173