Learn More
Thermo Scientific Chemicals Exendin (9-39)
For cellular and molecular biology applications
Supplier: Thermo Scientific Chemicals J66126EXD
Description
Exendin-3 (9-39) is a potent and selective GLP-1 receptor antagonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
3369.75 | |
0.5 mg |
-30°C to -10°C | |
Soluble in water at 1mg/ml | |
Exendin (9-39) | |
RUO - research use only |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only