Learn More
3M™ Exhalation Valve Filter 604
Exhalation valve filter is NIOSH-approved when used with 3M™ half facepiece 6000 series respirators.
Supplier: 3M™ 7100245392
Description
- Designed to help filter exhaled breath of respirator wearer
- Provides source control when worn with reusable respirator half facepiece 6000 series
- Easily installed and removed
- Users can conduct a positive pressure seal check while installed
- Hard case can be wiped with disinfecting wipes
Notes:
- Users should replace the exhalation valve filter if damaged, filter media becomes soiled, or if breathing becomes difficult
- NIOSH approved accessory only for use with 3M™ 6000 Series half facepiece respirators with 3M™ particulate filters 7093, 2091 or 5N11/501/603
- Not FDA cleared for use as a medical device for use in surgical applications
Specifications
ASTM F2101 | |
Clip | |
Exhalation Valve | |
604 |
NIOSH approved | |
Black | |
6000 Series half facepieces | |
30/Cs. |