Learn More
Welch™ Exhaust Filter with Drainback Feature
Description
- Exhaust can be directed into existing fume hood
- Reduces pump noise
- Use 1417E for pumps with NW16 Exhaust Connections (such as Welch CRVPro2, 4, 6, 8).
- Use 1417F for pumps with NW25 Exhaust Connections (such as Welch CRVpro16, 24, 30).
- Use 1417H-01 for replacement element for 1417F
Specifications
Specifications
| For Use With (Application) | Filters to 0.3 μm particles |
| For Use With | 1417F Exhaust Filters |
| Type | Replacement Element |