Learn More
Welch™ Exhaust Filter with Drainback Feature
Coalesces exhaust oil mist from rotary vane vacuum pump and drains back into oil case to reduce oil consumption and enable a clean work space. Exhaust can be directed into existing fume hood. Filters to 0.3 μm particles.
Supplier: Welch™ 1417F
Description
- Exhaust can be directed into existing fume hood
- Reduces pump noise
- Use 1417E for pumps with NW16 Exhaust Connections (such as Welch CRVPro2, 4, 6, 8).
- Use 1417F for pumps with NW25 Exhaust Connections (such as Welch CRVpro16, 24, 30).
- Use 1417H-01 for replacement element for 1417F
Specifications
Pumps with NW25 Exhaust Connections (Welch CRVpro16, 24, 30) | |
Filters to 0.3 μm particles | |
Exhaust Filter (Connection: NW25) |