Learn More
Welch™ Exhaust Filters for Welch™ DuoSeal™, Chemstar™ and Direct-Drive Pumps
Prevent damage to your pump by choosing to use an exhaust fulter from Welch for Duo Seal and Direct-Drive model pumps.
Manufacturer:  Welch™ 1417A
 View more versions of this product
Catalog No. 01-104-5B
Add to cart
Description
Description
Easily replaceable filter element captures oil mist from the exhaust port of the pump and reduces pump noise.
Specifications
Specifications
Exhaust Filter
Welch Pump Models 8907, 8912, 8917, 8920, 8925, 1376, 1376N, 1380, 1402, 1402N, 1405
Swivel Outlet with Exhaust Tubing Connector