Learn More
Welch™ Exhaust Filters for Welch™ DuoSeal™, Chemstar™ and Direct-Drive Pumps
Catalog No. p-159086
Change view
Click to view available options
Includes:
No Directional Outlet
Swivel Outlet, Exhaust Tubing Connector
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-104-5A No Directional Outlet
01-104-5B Swivel Outlet, Exhaust Tubing Connector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-104-5A Supplier Welch™ Supplier No. 1417
Add to Cart
Add to Cart
Prevent damage to your pump by choosing to use an exhaust fulter from Welch for Duo Seal and Direct-Drive model pumps.
Easily replaceable filter element captures oil mist from the exhaust port of the pump and reduces pump noise.
Specifications
For Use With Welch Pump Models 1399, 1400, 1400N, 8890, 8905
Includes No Directional Outlet
Type Exhaust Filter