Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

exo-2-Chloro-syn-7-hydroxymethyl-5-oxo-bicyclo[2.2.1]heptane 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
64768-55-4
C8H11ClO2
174.63
MFCD00161519
VOBDPILAUGFTKH-DBRKOABJSA-N
exo-2-chloro-syn-7-hydroxymethyl-5-oxo-bicyclo 2.2.1 heptane,1alpha,4alpha,7-anti-5alpha-chloro-7-hydroxymethyl bicyclo 2.2.1 heptane-2-one
51017543
(1R,4R,5R,7S)-5-chloro-7-(hydroxymethyl)bicyclo[2.2.1]heptan-2-one
C1C2C(C(C1Cl)CC2=O)CO
64768-55-4
C8H11ClO2
174.63
MFCD00161519
VOBDPILAUGFTKH-DBRKOABJSA-N
exo-2-chloro-syn-7-hydroxymethyl-5-oxo-bicyclo 2.2.1 heptane,1alpha,4alpha,7-anti-5alpha-chloro-7-hydroxymethyl bicyclo 2.2.1 heptane-2-one
51017543
(1R,4R,5R,7S)-5-chloro-7-(hydroxymethyl)bicyclo[2.2.1]heptan-2-one
C1C2C(C(C1Cl)CC2=O)CO
Melting Point 81.0°C to 85.0°C
Quantity 500mg
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 174.63
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material exo-2-Chloro-syn-7-hydroxymethyl-5-oxo-bicyclo[2.2.1]heptane, 98%