Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EXO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586470
Description
Purity is > 95% by SDS-PAGE.
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
Specifications
EXO1 | |
Polyclonal | |
Unconjugated | |
EXO1 | |
5730442G03Rik; Exo1; EXOI; Exonuclease 1; exonuclease I; HEX1; hExo1; hExoI; mExo1; Msa; rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26909, 305000, 9156 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9QZ11, Q9UQ84 | |
EXO1 | |
Synthetic peptide, corresponding to amino acids 55-05 of Human Exo1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction