Learn More
EXO1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 162531AP
Catalog No. 50-173-0831
Add to cart
Description
Description
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
Specifications
Specifications
EXO1
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9QZ11, Q9UQ84
EXO1, EXOI, exonuclease 1, Exonuclease I, HEX1, hExo1, hExoI
Rabbit
IgG
150 μL
-20°C
Primary
26909, 9156
Immunoprecipitation, Western Blot
Unconjugated
EXO1
Liquid
EXO1
EXO1 Fusion Protein Ag9253
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse