Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exo1, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
461681-88-9
C15H12FNO3
273.263
KIAPWMKFHIKQOZ-UHFFFAOYSA-N
exo1,methyl 2-4-fluorobenzoyl amino benzoate,exo-1,2-4-fluorobenzoylamino benzoic acid methyl ester,2-4-fluorobenzoylamino methylbenzoate,methyl 2-4-fluorobenzamido benzoate,exo 1,exo hplc , solid,methyl 2-4-fluorophenyl carbonylamino benzoate,2-4-fluoro-benzoylamino-benzoic acid methyl ester
310557
methyl 2-[(4-fluorobenzoyl)amino]benzoate
COC(=O)C1=CC=CC=C1NC(=O)C2=CC=C(C=C2)F
This item is not returnable. View return policy
461681-88-9
C15H12FNO3
273.263
KIAPWMKFHIKQOZ-UHFFFAOYSA-N
exo1,methyl 2-4-fluorobenzoyl amino benzoate,exo-1,2-4-fluorobenzoylamino benzoic acid methyl ester,2-4-fluorobenzoylamino methylbenzoate,methyl 2-4-fluorobenzamido benzoate,exo 1,exo hplc , solid,methyl 2-4-fluorophenyl carbonylamino benzoate,2-4-fluoro-benzoylamino-benzoic acid methyl ester
310557
methyl 2-[(4-fluorobenzoyl)amino]benzoate
COC(=O)C1=CC=CC=C1NC(=O)C2=CC=C(C=C2)F

Inhibits Golgi-ER traffic; blocks exocytosis

Reversible inhibitor of vesicular trafficking between endoplasmic reticulum and Golgi apparatus. Modifies Golgi ADP-ribosylation factor (ARF)1 GTPase activity and inhibits exocytosis (IC50 ∽ 20 mM). Has similar, but not identical, action to brefeldin A (Cat. No. 1231).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 273.26
Percent Purity >99%
Chemical Name or Material Exo1

Recommended Storage : Store at Room Temperature