Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exo1, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
461681-88-9
C15H12FNO3
273.263
KIAPWMKFHIKQOZ-UHFFFAOYSA-N
exo1,methyl 2-4-fluorobenzoyl amino benzoate,exo-1,2-4-fluorobenzoylamino benzoic acid methyl ester,2-4-fluorobenzoylamino methylbenzoate,methyl 2-4-fluorobenzamido benzoate,exo 1,exo hplc , solid,methyl 2-4-fluorophenyl carbonylamino benzoate,2-4-fluoro-benzoylamino-benzoic acid methyl ester
310557
methyl 2-[(4-fluorobenzoyl)amino]benzoate
COC(=O)C1=CC=CC=C1NC(=O)C2=CC=C(C=C2)F
This item is not returnable. View return policy
461681-88-9
C15H12FNO3
273.263
KIAPWMKFHIKQOZ-UHFFFAOYSA-N
exo1,methyl 2-4-fluorobenzoyl amino benzoate,exo-1,2-4-fluorobenzoylamino benzoic acid methyl ester,2-4-fluorobenzoylamino methylbenzoate,methyl 2-4-fluorobenzamido benzoate,exo 1,exo hplc , solid,methyl 2-4-fluorophenyl carbonylamino benzoate,2-4-fluoro-benzoylamino-benzoic acid methyl ester
310557
methyl 2-[(4-fluorobenzoyl)amino]benzoate
COC(=O)C1=CC=CC=C1NC(=O)C2=CC=C(C=C2)F

Inhibits Golgi-ER traffic; blocks exocytosis

Reversible inhibitor of vesicular trafficking between endoplasmic reticulum and Golgi apparatus. Modifies Golgi ADP-ribosylation factor (ARF)1 GTPase activity and inhibits exocytosis (IC 50 ~ 20 mM). Has similar, but not identical, action to brefeldin A (Cat. No. 1231).
Quantity 50mg
Formula Weight 273.26
Percent Purity >99%
Chemical Name or Material Exo1

Recommended Storage : Store at Room Temperature