Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC3 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI1D6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271881APCCY7

Catalog No. N27181APCY7


Add to cart

Description

Description

EXOSC3 Monoclonal antibody specifically detects EXOSC3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

EXOSC3
OTI1D6
Flow Cytometry
Affinity Purified, Conjugated
EXOSC3
Full length recombinant protein of human EXOSC3 (NP_057126) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
bA3J10.7, CGI-102, exosome complex exonuclease RRP40, exosome component 3MGC15120, exosome component Rrp40, hRrp-40, hRrp40p, p10Rrp40p, Ribosomal RNA-processing protein 40, RP11-3J10.8, RRP40MGC723
Mouse
IgG1
0.1 mL
Primary
51010.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit