Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome component 7 Mouse anti-Human, Mouse, Rat, Canine, Monkey, PE/Atto594, Clone: OTI1G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271722PEATT594

Catalog No. N271722P594


Add to cart

Description

Description

Exosome component 7 Monoclonal antibody specifically detects Exosome component 7 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Exosome component 7
OTI1G8
Flow Cytometry
Affinity Purified, Conjugated
EXOSC7
Full length recombinant protein of human Exosome component 7 (NP_055819) produced in HEK293T cells.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
23016.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit