Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exosome component 7 Rabbit anti-Human, Mouse, Clone: 6E5N8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Exosome component 7 |
---|---|
Clone | 6E5N8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Exosome component 7 Monoclonal antibody specifically detects Exosome component 7 in Human, Mouse samples. It is validated for Western BlotSpecifications
Exosome component 7 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
23016 | |
IgG | |
Affinity purified |
6E5N8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Exosome component 7 (Q15024). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title