Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EXP 3174

Potent and selective AT1 receptor antagonist

$143.00 - $600.00

Specifications

Chemical Name or Material 2-Butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid
Molecular Formula C22H21ClN6O2
CAS 124750-92-1
Product Type Target: Angiotensin AT1 Receptor Antagonists
Formula Weight 436.89
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-281-0 View Documents Tocris Bioscience™
5728/10
10mg Each for $143.00
Add to cart
 
57-285-0 View Documents Tocris Bioscience™
5728/50
50mg Each for $600.00
Add to cart
 
Description

Description

Potent and selective noncompetitive AT1 receptor antagonist (IC50 = 37 nM). Reduces blood pressure in a rat model. Active metabolite of losartan potassium (Cat.No. 3798).
Specifications

Specifications

2-Butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid
124750-92-1
436.89
C22H21ClN6O2
Target: Angiotensin AT1 Receptor Antagonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit