Learn More
Research Products International CorpSupplier Diversity Partner Explorer Semi-Micro Balance, Ohaus EX225/AD, 220 gm  DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 400900
Intelligent PERFORMANCE – All Explorers have newly optimized linearity and repeatability specifications and enhanced vibration filtering for better balance stability. All models also come standard with AutoCal automatic internal calibration. 3×3 filtering setup ensures fast and stable weighing results. models offer traditional external calibration. These combined enhancements improve accuracy, efficiency, and throughput.
Catalog No. 50-197-6800