Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exploring with a Microscope Products for Science Education

Study a whole microworld.

Specifications

Class Size 40 students
For Use With (Application) Study a whole microworld
View More Specs

Includes:
  • Magnifiers
  • Nut and bolt microtomes
  • Paraffin
  • Pipets
  • Polarizing filters
  • Pond mix
  • Rulers
  • Slides
  • Stains
  • Teacher and student guides

Requires: Compound microscopes


Products 1
Catalog Number Mfr. No. Includes Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Includes Product Type Price Quantity & Availability  
S63072 Products for Science Education
Required Accessories View Documents
Neo/Sci Corporation
201063
Magnifiers, nut and bolt microtomes, paraffin, pipets, polarizing filters, pond mix, rulers, slides, stains, teacher and student guides Microscope Kit This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Prepare slides and explore the microworld of plants, animals and microbes. Construct a water drop microscope and learn new microscope imaging techniques — such as darkfield illumination and polarizing light — to obtain a unique view of the “World of Little.”

Order Info

Class size: 40 students.

Specifications

Specifications

40 students
Study a whole microworld
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit