Explosion-Proof Freezers

Maintain a safe work environment even in hazardous conditions with this explosion-proof freezer.

Manufacturer: Thermo Scientific 20EFEETSV

 View more versions of this product

Catalog No. 20EFEETSV

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Dimensions (L x W x H) Exterior 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 185.7 cm)
    Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
    Temperature Range (Metric) -12°C to -20°C
    Color White
    Control Mechanical Thermostat/Outside, On Top Of Unit, In Blue Housing
    Warranty 13 Months Parts and Labor
    Current 5 Amps/1150 Watts
    Casters Adjustable Leveling Feet
    Compressor Bottom Mounted
    Exterior Finish Epoxy Coated Steel
    Interior Finish Epoxy Coated Steel
    Model 20EFEETSV
    Shelves 4 Fixed Shelves, 6 Fixed Door Shelves
    Type Explosion Proof Freezer
    Cabinet Material Enameled steel, White,Enameled steel, White
    Voltage 230V
    Doors Single (one compartment)
    Certifications/Compliance UL listed meets NFPA 45 and 70
    Door Count 1 (1 compartment),1(1 compartment)
    Height (English) Exterior 67.25"
    Hertz 50Hz,50Hz
    Length (Metric) Exterior 79.4cm
    Amperage 5.0A,5.0A
    Plug Type No Cord Supplied
    Capacity 20.0 cu. ft. / 566.34L,20.9 Cubic Feet/591.82 Liters
    Refrigerant R134A,CFCFree,R134a,100% CFC Free
    Width (English) Exterior 33 in.
    Width (Metric) Exterior 83.8cm
    Capacity (English) 20.0 cu. ft.
    Chamber Material Enameled steel,Enameled steel
    Defrost Manual Defrost
    Insulation Polyurethane Foam,Polyurethane Foam
    Capacity (Metric) 566.34L
    Door Style Hinged, Opens Left to Right, Non-Reversible,Hinged, Opens Left to Right, Non-Reversible
    Shipping Weight (Metric) 128kg
    Depth (Metric) Interior 53.34cm
    Electrical Requirements 230 V/50 Hz
    Height (Metric) Interior 68.58cm
    Shipping Weight (English) 230 lb.
    Width (Metric) Interior 68.58cm
    Height (English) Interior 57.5"
    Height (Metric) Exterior 170.8cm
    Length (English) Exterior 31.25 in.
    Width (English) Interior 27 in.
    Depth (English) Interior 21"

    Thermo Scientific explosion-proof laboratory refrigerators, freezers and combinations units are particularly suited for safe storage in hazardous environments.

    Features:

    • Temperature range: -20° to -12°C
    • Thermostat and compressor components isolated within a vapor-proof enclosure
    • Electrical connections hardwired to prevent sparking and accidental disconnection
    • Magnetic vinyl gaskets for positive door seal
    • CFC-free insulation and refrigerant
    • Hydraulic thermostat
    • Two fixed shelves, three door shelves
    • Aluminum evaporator coil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit