Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Explosion-Proof Freezers Available on GSA/VA Contract for Federal Government customers only.

Maintain a safe work environment even in hazaRDO™us conditions with this explosion-proof freezer.

$5110.00

Specifications

Amperage 1.1 A
Cabinet Material Epoxy-enameled Steel, White
Certifications/Compliance NFPA 45 and 70, UL
Defrost Manual
Shipping Weight (English) 120 lb.
View More Specs
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Products 2
Catalog Number Mfr. No. Capacity (Metric) Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Electrical Requirements Price Quantity & Availability  
05EFEETSA Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
05EFEETSA
141.6 L 115 V 60 Hz
Each for $5,110.00
Add to cart
 
20EFEETSA
Accessories View Documents
Thermo Scientific™
20EFEETSA
566.34 L 115 V 60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

Use Thermo Scientific™ explosion-proof laboratory refrigerators, freezers and combination units for safe storage in hazaRDO™us environments.

Features:
  • Temperature range: -20° to -12°C
  • Thermostat and compressor components isolated within a vapor-proof enclosure
  • Electrical connections hardwired to prevent sparking and accidental disconnection
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
  • Hydraulic thermostat
  • Two fixed shelves, three door shelves
  • Aluminum evaporator coil
  • Warranty varies by country, please contact us for details

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Warranty and Services

13 Months Parts and Labor

Specifications

Specifications

1.1 A
NFPA 45 and 70, UL
120 lb.
1°C to 12°C
Polyurethane Foam
Epoxy-enameled Steel, White
Manual
54.49 kg
Hinged, Open Left to Right, Non-Reversible
R134a, 100% CFC Free
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit