Thermo Scientific™ Explosion-Proof Refrigerator

Maintain a safe work environment with Thermo Scientific™ Explosion-Proof Refrigerators, engineered to prevent triggering an explosion inside or outside the unit.

Manufacturer: Thermo Scientific™ 20EREETSA

 View more versions of this product

Catalog No. 20EREETSA

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
    Typical Lifetime 10 Years
    Min. Clearance 3 in. On Sides, 3 in. At Back, 3 in. On Top
    Lighting Interior - none
    Temperature (Setpoint) Adjustable Set Point - Yes
    Display Temperature Display - None
    Dimensions (L x W x H) Exterior 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1cm)
    Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
    Shelves 4 fixed Interior shelves, 1 Wire Basket, 6 door shelves,4 Fixed Shelves, 6 Fixed Door Shelves
    Voltage 115V,120V
    Certifications/Compliance UL listed meets NFPA 45 and 70
    Electrical Requirements 115 V/60 Hz
    Color White
    Control Mechanical / Back of the unit
    Warranty 13 Months Parts and Labor
    Description Explosion-Proof Refrigerator
    Current 5 Amps/575 Watts
    Amperage 5.0A
    Capacity 20.0 cu. ft. / 566.34L,20.9 Cubic Feet/591.82 Liters,20.0 cu. ft. / 566.34L
    Capacity (English) 20.0 cu. ft.
    Capacity (Metric) 566.34L
    Casters 2 Adjustable Feet In Front, 2 Fixed Pads In Rear,Adjustable Leveling Feet
    Defrost Manual,Manual Defrost
    Compressor Bottom Mounted
    Door Count 1 (1 compartment),1(1 compartment)
    Door Style Hinged, Opens Left to Right, Non-Reversible
    Exterior Finish Enameled Steel,Epoxy Coated Steel
    Hertz 60Hz
    Interior Finish ABS Plastic,Epoxy Coated Steel
    Insulation Polyurethane Foam,Polyurethane Foam
    Model 20EREETSA
    Access Security Door Lock
    Plug Type No Cord Supplied
    Shipping Weight (English) 230 lb.,283 lb.
    Refrigerant R134A, CFCFree,R134a,100% CFC Free,R134A,CFCFree
    Shipping Weight (Metric) 104.3kg,128kg
    Temperature Range (Metric) 1° to 12°C,1°C to 12°C,1° to 12°C
    Type Explosion Proof Refrigerator
    Height (English) Exterior 67.25 in.
    Height (English) Interior 57.5 in.
    Height (Metric) Interior 146.05cm
    Depth (Metric) Interior 53.34cm
    Height (Metric) Exterior 170.8cm
    Width (English) Interior 27 in.
    Cabinet Material Epoxy-enameled steel, White
    Width (Metric) Exterior 83.8cm
    Depth (English) Interior 21 in.
    Length (English) Exterior 31.25 in.
    Length (Metric) Exterior 79.4cm
    Width (English) Exterior 33 in.
    Width (Metric) Interior 68.58cm
    Chamber Material Epoxy Enameled Steel

    Prevent the triggering of an explosion inside or outside the unit with Thermo Scientific™ Explosion-Proof Refrigerators, suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.

    • Hydraulic thermostat
    • Manual defrost
    • Magnetic vinyl gaskets for positive door seal
    • CFC-free insulation and refrigerant

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit