Explosion-Proof Refrigerator

Maintain a safe work environment with Thermo Scientific™ Explosion-Proof Refrigerators, engineered to prevent triggering an explosion inside or outside the unit.

Manufacturer: Thermo Scientific 20ERCETSA

 View more versions of this product

Catalog No. 20ERCETSA

  Check Availability
Add to Cart

    Specifications

    Dimensions (L x W x H) Exterior 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1cm)
    Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
    Temperature Range (Metric) 1°C to 12°C
    Min. Clearance 3 in. On Sides, 3 in. At Back, 3 in. On Top
    For Use With (Application) Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
    Typical Lifetime 10 Years
    Lighting Interior - none
    Temperature (Setpoint) Adjustable Set Point - Yes
    Display Temperature Display - None
    Warranty 13 Months Parts and Labor
    Model 20ERCETSA
    Type Explosion Proof Refrigerator
    Voltage 115V
    Certifications/Compliance cCSAus
    Electrical Requirements 115 V/60 Hz
    Color White
    Control Mechanical Thermostat/Outside, On Top Of Unit, In Blue Housing
    Description Corrosion Resistant, Explosion-Proof Refrigerator
    Current 5 Amps/575 Watts
    Capacity 20.9 Cubic Feet/591.82 Liters,20.0 cu. ft. / 566.34L
    Capacity (English) 20.0 cu. ft.
    Capacity (Metric) 566.34L
    Casters Adjustable Leveling Feet
    Compressor Bottom Mounted
    Door Count 1(1 compartment)
    Door Style Hinged, Open Left to Right, Non-Reversible
    Exterior Finish Epoxy Coated Steel
    Hertz 60Hz
    Interior Finish Epoxy Coated Steel
    Insulation Polyurethane Foam,Polyurethane Foam
    Access Security Door Lock
    Plug Type No Cord Supplied
    Shelves 4 Fixed Shelves
    Refrigerant R134a,100% CFC Free
    Shipping Weight (Metric) 104.3kg
    Amperage 5.0A
    Depth (Metric) Interior 53.34cm
    Height (English) Exterior 67.25 in.
    Height (English) Interior 57.5 in.
    Height (Metric) Exterior 170.8cm
    Height (Metric) Interior 146.05cm
    Length (English) Exterior 31.25 in.
    Length (Metric) Exterior 79.4cm
    Shipping Weight (English) 230 lb.
    Width (English) Exterior 33 in.
    Width (English) Interior 27 in.
    Cabinet Material Epoxy-enameled steel, White
    Width (Metric) Exterior 83.8cm
    Width (Metric) Interior 68.58cm
    Defrost Manual Defrost
    Chamber Material Epoxy Enameled Steel
    Depth (English) Interior 21 in.

    Prevent the triggering of an explosion inside or outside the unit with Thermo Scientific™ Explosion-Proof Refrigerators, suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.

    • Hydraulic thermostat
    • Manual defrost
    • Magnetic vinyl gaskets for positive door seal
    • CFC-free insulation and refrigerant

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit