Learn More
Thermo Scientific™ Explosion-Proof Refrigerator
Maintain a safe work environment with Thermo Scientific™ Explosion-Proof Refrigerators, engineered to prevent triggering an explosion inside or outside the unit.
Manufacturer:  Thermo Scientific™ 20ERCETSV
 View more versions of this product
Catalog No. 20ERCETSV
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications
Description
Prevent the triggering of an explosion inside or outside the unit with Thermo Scientific™ Explosion-Proof Refrigerators, suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.
  • Hydraulic thermostat
  • Manual defrost
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
Description & Specifications
Specifications
Door Lock
Mechanical Thermostat/Outside, On Top Of Unit, In Blue Housing
2.5Amps/600 Watts
20.9 Cubic Feet/591.82 Liters
566.34L
Manual Defrost
Temperature Display - None
Hinged, Open Left to Right, Non-Reversible
32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1cm)
50Hz
Polyurethane Foam
No Cord Supplied
4 Fixed Shelves
104.3kg
Explosion Proof Refrigerator
230 V/50 Hz
Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
3 in. On Sides, 3 in. At Back, 3 in. On Top
Adjustable Set Point - Yes
White
Corrosion Resistant, Explosion-Proof Refrigerator
2.5A
20.0 cu. ft.
Adjustable Leveling Feet
Bottom Mounted
1(1 compartment)
Epoxy Coated Steel
23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
Epoxy Coated Steel
20ERCETSV
230 lb.
R134a,100% CFC Free
1°C to 12°C
230V
Warranty varies by country, please contact us for details
10 Years
Interior - none