Thermo Scientific™ Explosion-Proof Refrigerator

Maintain a safe work environment with Thermo Scientific™ Explosion-Proof Refrigerators, engineered to prevent triggering an explosion inside or outside the unit.

Manufacturer: Thermo Scientific™ 20EREETSV

 View more versions of this product

Catalog No. 20EREETSV

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Dimensions (L x W x H) Exterior 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1cm)
    Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
    Temperature Range (Metric) 1°C to 12°C
    Min. Clearance 3 in. On Sides, 3 in. At Back, 3 in. On Top
    For Use With (Application) Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
    Temperature (Setpoint) Adjustable Set Point - Yes
    Display Temperature Display - None
    Typical Lifetime 10 Years
    Lighting Interior - none
    Warranty 13 Months Parts and Labor
    Current 2.0Amps/186 Watts
    Model 20EREETSV
    Type Explosion Proof Refrigerator
    Voltage 230V
    Certifications/Compliance UL listed meets NFPA 45 and 70
    Color White
    Control Mechanical Thermostat/Outside, On Top Of Unit, In Blue Housing
    Amperage 2.5A
    Capacity (Metric) 566.34L
    Casters Adjustable Leveling Feet
    Defrost Manual Defrost
    Compressor Bottom Mounted
    Door Style Hinged, Opens Left to Right, Non-Reversible
    Exterior Finish Epoxy Coated Steel
    Interior Finish Epoxy Coated Steel
    Insulation Polyurethane Foam,Polyurethane Foam
    Access Security Door Lock
    Shipping Weight (English) 283 lb.
    Shelves 4 Fixed Shelves, 6 Fixed Door Shelves
    Refrigerant R134a,100% CFC Free
    Depth (Metric) Interior 53.34cm
    Electrical Requirements 230 V/50 Hz
    Door Count 1 (1 compartment)
    Height (English) Exterior 67.25 in.
    Height (English) Interior 57.5 in.
    Description Explosion-Proof Refrigerator
    Height (Metric) Interior 146.05cm
    Hertz 50Hz
    Length (English) Exterior 31.25 in.
    Length (Metric) Exterior 79.4cm
    Plug Type No Cord Supplied
    Capacity 20.9 Cubic Feet/591.82 Liters,20.0 cu. ft. / 566.34L
    Shipping Weight (Metric) 104.3kg
    Capacity (English) 20.0 cu. ft.
    Width (English) Interior 27 in.
    Cabinet Material Epoxy-enameled steel, White
    Width (Metric) Exterior 83.8cm
    Width (Metric) Interior 68.58cm
    Chamber Material Epoxy Enameled Steel
    Depth (English) Interior 21 in.
    Height (Metric) Exterior 170.8cm
    Width (English) Exterior 33 in.

    Prevent the triggering of an explosion inside or outside the unit with Thermo Scientific™ Explosion-Proof Refrigerators, suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit