Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exportin-5 Rat anti-Human, Clone: EX5 4E1, MilliporeSigma™

Rat Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen Exportin-5
Clone EX5 4E1
Concentration 0.5 mg/mL
Applications ELISA, Immunofluorescence, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE1922100
View Documents
MilliporeSigma
MABE1922100UL
100 μL
Each for $373.00
Add to cart
 
MABE192225
View Documents
MilliporeSigma
MABE192225UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

Exportin-5 Monoclonal antibody specifically detects Exportin-5 in Human samples. It is validated for ELISA, Immunofluorescence, Western Blotting.
Specifications

Specifications

Exportin-5
0.5 mg/mL
Unconjugated
Rat
Human
Ovalbumin-conjugated linear peptide corresponding to 15 amino acids from the N-terminal half of human Exportin-5.
Monoclonal
Protein G purified
EX5 4E1
ELISA, Immunofluorescence, Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit