Learn More
Chemglass Life Sciences 10ft Extension Cord, Type "T" (Blue), Standard to Mini Plug
SDP
Supplier:  Chemglass Life Sciences CG349902
Encompass_Preferred
10ft Extension Cord, Type "T" (Blue), Standard to Mini Plug. Coiled extension cords to connect thermocouples to instrumentation such as temperature monitors, controllers and data loggers. Cords are 10ft long and made of 26-gauge multistrand wire with PVC insulation. All cords have a female standard (OST) with a choice of a male standard (OST) or mini (SMP) connection.
Catalog No. CG349902
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.