Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Extension Collar for CoolBox™ 2XT System
For use with CoolBox™ 2XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.
Supplier: Corning™ 432088
Alert:
Lid closure not possible with addition of extension collar
Description
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.Specifications
Extension Collar | |
1/Pk. | |
Extend the height when using tall tube modules | |
Interior: 21.0 x 14.3 x 7.0 cm (8.2 x 5.6 x 2.7 in.) |
Green | |
1/Cs. | |
CoolBox 2XT System | |
26.7 x 20.0 x 7.0 cm (6.5 x 7.8 x 2.7 in.) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction