Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Extensor Long PCR Enzyme Mix Products

Amplify long PCR products up to 20kb with this synergistically-acting PCR enzyme blend of Taq DNA polymerase and a proprietary proofreading enzyme.

Specifications

For Use With (Application) Amplifying fragments up to 20kb; PCR; Cloning; Long PCR
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles
View More Specs

Products 2
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
AB-0792B
View Documents
Thermo Scientific™
AB0792B
Extensor Master Mix (Buffer 1 Formulation) 500 25µL Rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
AB-0794B
View Documents
Thermo Scientific™
AB0794B
Extensor ReddyMix Master Mix (Buffer 1 Formulation) 400 x 25µL Rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Extensor Long Range PCR Enzyme is a blend of ThermoPrime Taq DNA Polymerase and a proprietary thermostable proofreading enzyme. The two enzymes act synergistically to generate long PCR products, up to 20 kb. Extensor Long Range Enzyme includes 2 buffer formulations; Buffer 1 for templates up to 12 kb in length, and Buffer 2 for templates 12 to 20 kb and GC-rich or other difficult templates. This enzyme blend is also available in 2X Master Mix and ReddyMix formulations. The ReddyMix Master Mix contains an inert red dye and a density reagent that allows for direct loading onto agarose gels.

Highlights

  • Proprietary enzyme blend amplifies targets up to 20 kb in length
  • Alternative buffering system for success with GC rich targets
Specifications

Specifications

Amplifying fragments up to 20kb; PCR; Cloning; Long PCR
Store at -20°C. Avoid freeze/thaw cycles

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit