Learn More
Brady™ Extra Large Lockout Station with Steel Locks
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Extra Large Lockout Station with Steel Locks is constructed of sturdy white polystyrene with predrilled holes for easy wall mounting. Pegboard-style holes for hooks drilled into back (8 hooks supplied standard).
Manufacturer:  Brady™ 99694
Catalog No. 19-104-028
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Text Legend: LOCKOUT STATION
30 in.
5.25 in.
25 in.
Safety
Red on white
(18) 0.75 in shackle red kd steel padlock; (2) 1 to 2.5 in gate valve lockout; (2) 277 v lockout cleat; (2) 277 v no-hole circuit breaker; (2) 480/600 v lockout cleat; (2) 480/600 v no-hole circuit breaker; (2) single hole circuit breaker lockout; (25) lockout tag; 1 to 6.5 in adjustable gate valve lockout; 110 v plug lockout; 3-in-1 plug lockout; all purpose cable lockout; large ball valve lockout; oversized circuit breaker lockout; small ball valve lockout; universal valve lockout; wall switch lockout
Polystyrene
5.25 in. x 25 in. x 30 in.