Learn More
ValueMax™ Extra-Safe™ Lab Coats — Teal
Static-free — no clinging
Supplier: ValueMax™ 3660TE4XL
This item is not returnable.
View return policy
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
- Fluid-resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Knee length
- Autoclavable and washable up to five times
- Latex-free, fire retardant
Specifications
4X-Large | |
3-ply SMS | |
Snaps | |
Knee length | |
Yes |
SMS | |
Teal | |
Yes | |
3 | |
3-ply |