Learn More
ValueMax™ Extra-Safe™ Lab Coats — Teal
Static-free — no clinging
Catalog Number | Mfr. No. | Material | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Size | Price | Quantity & Availability | ||||
23-900-511D
|
ValueMax™
3660TEXL |
Polypropylene | X-Large | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
23-900-511G
|
ValueMax™
3660TE4XL |
SMS | 4X-Large | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
23-900-511B
|
ValueMax™
3660TEM |
Polypropylene | Medium | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
23-900-511C
|
ValueMax™
3660TEL |
Polypropylene | Large | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
23-900-511E
|
ValueMax™
3660TE2XL |
Polypropylene | 2X-Large | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
- Fluid-resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Knee length
- Autoclavable and washable up to five times
- Latex-free, fire retardant
Specifications
Teal | |
3 | |
Yes |