Learn More
Apex Aseptic Extra-Safe Lab Jackets Medical Blue
Static-free no clinging
Manufacturer: Apex Aseptic 3630MB5XL
Description
- Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
- Fluid- resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Hip length
- Autoclavable and washable up to five times
- Latex- free, fire retardant
Specifications
Knit | |
Snaps | |
Knit | |
3 | |
Yes |
OSHA | |
Medical Blue | |
SMS | |
5X-Large | |
3-ply |