Learn More
Air Impurities Removal Systems Extract-ALL™ Filters for Bench-Top Clean Air Station
Manufacturer:  Air Impurities Removal Systems F9845A
 View more versions of this product
Catalog No. 19-097-455
Description
Description
Specifications
Specifications
Filter
60%
Captures solvent, touch-up paint, spray glue fumes and other gaseous odors
Carbon
Benchtop ductless clean air station