Extract-ALL™ Portable Fume Extraction System
Manufacturer: Air Impurities Removal Systems, IncSupplier Diversity Partner S9872
 View more versions of this product
Catalog No. 19-097-415
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance CE and UL-approved blower
    Electrical Requirements 120V 60Hz, 200W
    For Use With (Application) For removing contaminants caused by chemical mixing, soldering, sanding, grinding, light welding, solvents and other light to moderate industrial or laboratory activities
    Voltage 120V
    Type Portable Fume Extraction System
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback